Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethyl orthoformate

Catalog No. 10845620L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-845-620L 20 L
10-845-6500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-845-620L Supplier Sigma Aldrich Supplier No. 10845620L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Trimethyl orthoformate is an effective solvent for thallium(iii) nitrate-mediated oxidations
  • It undergoes acid catalyzed reaction with 6-(n-d-ribitylanilino) uracils to form 8-demethyl-8-hydroxy-5-deazariboflavins.trimethyl orthoformate was used as dehydrating agent in the preparation of surface-modified colloidal silica nanoparticles

Specifications

CAS 149-73-5
Density 0.97 g/mL (at 25°C (literature))
Boiling Point 101°C to 102°C (lit.)
Molecular Formula C4H10O3
Refractive Index n20/D 1.379 (literature)
Linear Formula CH(OCH3)3
MDL Number MFCD00008483
Quantity 20 L
Synonym Trimethoxymethane
Molecular Weight (g/mol) 106.12
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.