Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl Orthoisobutyrate 98.0+%, TCI America™
SDP

Catalog No. T220825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T220825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T220825G Supplier TCI America Supplier No. T220825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52698-46-1
Molecular Formula C7H16O3
Molecular Weight (g/mol) 148.20
MDL Number MFCD08276428
InChI Key BGLARIMANCDMQX-UHFFFAOYSA-N
Synonym Orthoisobutyric Acid Trimethyl Ester, 1,1,1-Trimethoxy-2-methylpropane
PubChem CID 40495
IUPAC Name 1,1,1-trimethoxy-2-methylpropane
SMILES COC(OC)(OC)C(C)C

Specifications

Color Colorless
Quantity 25 g
UN Number 3272
Formula Weight 148.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl Orthoisobutyrate
EINECSNumber (2)-3640
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.