Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Trimethyl-p-tolylsilane, 97%

Click to view available options
Quantity:
5g
25g
100g
3728-43-6
C10H16Si
164.323
MFCD00014850
QGHURGPPCGMAMZ-UHFFFAOYSA-N
trimethyl-p-tolylsilane,p-tolyltrimethylsilane,trimethyl 4-methylphenyl silane,trimethyl-p-tolyl silane,trimethyl p-tolyl silane,4-trimethylsilyl toluene,silane, p-tolyl-trimethyl-,,4-methylphenyl trimethylsilane
298149
trimethyl-(4-methylphenyl)silane
CC1=CC=C(C=C1)[Si](C)(C)C
3728-43-6
C10H16Si
164.323
MFCD00014850
QGHURGPPCGMAMZ-UHFFFAOYSA-N
trimethyl-p-tolylsilane,p-tolyltrimethylsilane,trimethyl 4-methylphenyl silane,trimethyl-p-tolyl silane,trimethyl p-tolyl silane,4-trimethylsilyl toluene,silane, p-tolyl-trimethyl-,,4-methylphenyl trimethylsilane
298149
trimethyl-(4-methylphenyl)silane
CC1=CC=C(C=C1)[Si](C)(C)C
Applications
It is employed in the production of trimethyl-(4-trimethylsilanyl-benzyl)-silane by reacting with chloro-trimethyl-silane.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect against electrostatic charges. Keep ignition sources away.
UN Number UN1993
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 164.33
Percent Purity 97%
Density 0.865
Refractive Index 1.491
Boiling Point 191°C to 192°C
Quantity 25g
Flash Point 46°C (115°F)
Chemical Name or Material Trimethyl-p-tolylsilane

GHS H Statement
H226-H315-H319
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures