Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl phosphate, 98+%

Catalog No. AAA1059536
Click to view available options
Quantity:
500 g
2500 g

CAS: 512-56-1 | C3H9O4P | 140.08 g/mol

Trimethyl phosphate is used as a color inhibitor for polyester and other polymer preparations, flame retardants and plasticizer. It acts as a solvent for aromatic halogenations and nitration reactions. It is used as a mild methylating reagent which is used in the dimethylation of aniline and related heterocyclic compound. Furthermore, it is involved in the synthesis of 1-methyl-pyridinium; dimethyl phosphate by reacting with pyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethyl phosphate is used as a color inhibitor for polyester and other polymer preparations, flame retardants and plasticizer. It acts as a solvent for aromatic halogenations and nitration reactions. It is used as a mild methylating reagent which is used in the dimethylation of aniline and related heterocyclic compound. Furthermore, it is involved in the synthesis of 1-methyl-pyridinium; dimethyl phosphate by reacting with pyridine.

Solubility
Miscible with water.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 512-56-1
Molecular Formula C3H9O4P
Molecular Weight (g/mol) 140.08
MDL Number MFCD00008348
InChI Key WVLBCYQITXONBZ-UHFFFAOYSA-N
Synonym trimethylphosphate, phosphoric acid, trimethyl ester, trimethyl orthophosphate, tmpo, trimethoxyphosphine oxide, phosphoric acid trimethyl ester, o,o,o-trimethyl phosphate, trimethylfosfat, phosphate, trimethyl, trimethylfosfat czech
PubChem CID 10541
ChEBI CHEBI:46324
IUPAC Name trimethyl phosphate
SMILES COP(=O)(OC)OC

Specifications

Melting Point -46°C
Density 1.213
Boiling Point 196°C to 198°C
Flash Point >150°C (302°F)
Odor Pleasant
Refractive Index 1.397
Linear Formula (CH3O)3PO
Quantity 500 g
Beilstein 1071731
Sensitivity Moisture sensitive
Solubility Information Miscible with water.
Formula Weight 140.08
Percent Purity ≥98%
Chemical Name or Material Trimethyl phosphate
Show More Show Less
missing translation for 'healthHazard1' H302-H315-H319-H340-H351
missing translation for 'healthHazard2' GHS H Statement
H340-H351-H302-H315-H319
May cause genetic defects.
Suspected of causing cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
missing translation for 'healthHazard3' P201-P202-P264b-P270-P280i-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
EINECSNumber 208-144-8
RTECSNumber TC8225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.