Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl phosphate, 99%

Catalog No. AC157970025
Change view
Click to view available options
Quantity:
50 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157970025 2.5 L Glass bottle
AC157970500 50 mL Glass bottle
AC157975000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC157970025 Supplier Thermo Scientific Chemicals Supplier No. 157970025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 512-56-1 | C3H9O4P | 140.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 512-56-1
Molecular Formula C3H9O4P
Molecular Weight (g/mol) 140.08
MDL Number MFCD00008348
InChI Key WVLBCYQITXONBZ-UHFFFAOYSA-N
Synonym trimethylphosphate, phosphoric acid, trimethyl ester, trimethyl orthophosphate, tmpo, trimethoxyphosphine oxide, phosphoric acid trimethyl ester, o,o,o-trimethyl phosphate, trimethylfosfat, phosphate, trimethyl, trimethylfosfat czech
PubChem CID 10541
ChEBI CHEBI:46324
IUPAC Name trimethyl phosphate
SMILES COP(=O)(OC)OC

Specifications

Melting Point -46.0°C
Density 1.1970g/mL
Boiling Point 197.0°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3950 to 1.3970
Linear Formula (CH3O)3P(O)
Quantity 2.5 L
Beilstein 01,286
Fieser 05,716; 07,393
Specific Gravity 1.197
Solubility Information Solubility in water: 500g/L (25°C). Other solubilities: soluble in gasoline,soluble in alcohol,ether
Formula Weight 140.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Trimethyl phosphate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause genetic defects.
Suspected of causing cancer.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
EINECSNumber 208-144-8
RTECSNumber TC8225000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.