Learn More
Sigma Organic Chemistry Trimethyl phosphite | 50ML | 121-45-9 | MFCD00008350
Supplier: Sigma Organic Chemistry 9273050ML

Trimethyl phosphite, 50ML
About This Item:
Synonym(s): TMP; Methyl phosphite; P(OMe)3; Trimethoxyphosphine
Linear Formula: (CH3O)3P
Molecular Weight: 124.08
CAS Number: 121-45-9
MDL Number: MFCD00008350
UNSPSC Code: 12350000
Purity: >=97%
Boiling Point: 111-112 C (lit.)
Melting Point: -78 C (lit.)
Density: 1.052 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.408 (lit.); n20/D 1.408
Storage: 2-8C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.