Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylacetic anhydride, 99%

Catalog No. AC162440500
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC162440500 50 mL Glass bottle
AC162442500 250 mL Glass bottle
AC162440010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC162440500 Supplier Thermo Scientific Chemicals Supplier No. 162440500
Add to Cart

CAS: 1538-75-6 | C10H18O3 | 186.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1538-75-6
Molecular Formula C10H18O3
Molecular Weight (g/mol) 186.25
MDL Number MFCD00008842
InChI Key PGZVFRAEAAXREB-UHFFFAOYSA-N
Synonym pivalic anhydride, trimethylacetic anhydride, 2,2-dimethylpropionic anhydride, propanoic acid, 2,2-dimethyl-, anhydride, pivalic anydride, 2,2-dimethylpropanoic anhydride, trimethylacetic acid anhydride, propanoic acid, 2,2-dimethyl-, 1,1'-anhydride, trimethylaceticanhydride, bispivalic anhydride
PubChem CID 15234
IUPAC Name 2,2-dimethylpropanoyl 2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)OC(=O)C(C)(C)C

Specifications

Color Colorless
Density 0.9100g/mL
Boiling Point 193.0°C
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4080 to 1.4100
Linear Formula [(CH3)3CCO]2O
Quantity 50 mL
Beilstein 02,32
Specific Gravity 0.91
Solubility Information Solubility in water: decomposes in water
Formula Weight 186.25
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Trimethylacetic anhydride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 216-263-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.