Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylaluminum, 25% w/w in hexane, Thermo Scientific™

Catalog No. AA42836AP
Change view
Click to view available options
Quantity:
500 mL
4 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42836AP 500 mL
AA42836AY 4 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42836AP Supplier Thermo Scientific Chemicals Supplier No. 42836AP

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-24-1
Molecular Formula C3H9Al
Molecular Weight (g/mol) 72.087
MDL Number MFCD00008252
InChI Key JLTRXTDYQLMHGR-UHFFFAOYSA-N
Synonym trimethylaluminum, trimethylaluminium, aluminum, trimethyl, trimethylalane, unii-av210lg46j, ch3 3al, aluminum trimethanide, aluminum trimethyl, trimethylaluminum solution, trimethylaluminum, elec. gr.
PubChem CID 16682925
IUPAC Name trimethylalumane
SMILES C[Al](C)C

Specifications

Flash Point −18°C (−1°F)
Quantity 500 mL
UN Number UN3399
Beilstein 3587197
Sensitivity Air and moisture sensitive
Solubility Information Soluble in aromatic,saturated aliphatic and cycloaliphatic hydrocarbons.
Formula Weight 72.09
Physical Form Liquid
Chemical Name or Material Trimethylaluminum
Hazard Category H225-H250-H260-H304-H314-H335-H336-H361f-H373
Hazard Statement GHS H Statement
H225-H260-H304-H361-H373-H314-H336
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
EINECSNumber 200-853-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.