Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylaluminum Solution

Catalog No. 268569100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-856-9100ML 100 mL
26-856-9800ML 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 26-856-9100ML Supplier Sigma Aldrich Supplier No. 268569100ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethylaluminum is a Lewis acid having wide applications in organic synthesis. It is employed in organic reactions like cross-coupling reaction, methylenation, carboalumination, cyclopropanation, and rearrangement reactions. Tebbe Reagent (methylenation agent) can be prepared in situ by reacting trimethylaluminum with dichlorobis(cyclopentadienyl)titanium.Used with 4-bromo-2,6-di-tert-butylphenol (cat. no. 394378) to prepare a wide range of carbonyl compounds via the selective rearrangement of epoxides.

Specifications

CAS 75-24-1
Density 0.683 g/mL at 25°C
Molecular Formula C3H9Al
Linear Formula (CH3)3Al
MDL Number MFCD00008252
Quantity 100 mL
Molecular Weight (g/mol) 72.09 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.