Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylamine, 33% w/w in ethanol denatured with 2% cyclohexane

Catalog No. AAH27324AK
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27324AK 250 mL
AAH27324AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27324AK Supplier Thermo Scientific Chemicals Supplier No. H27324AK
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 75-50-3 | C3H9N | 59.11 g/mol

Mainly as intermediate used in the production of industrial chemicals, initial product for chemical synthesis, dyes, textile industry, active ingredient. Trimethylamine is used in the synthesis of choline, tetramethylammonium hydroxide, plant growth regulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mainly as intermediate used in the production of industrial chemicals, initial product for chemical synthesis, dyes, textile industry, active ingredient. Trimethylamine is used in the synthesis of choline, tetramethylammonium hydroxide, plant growth regulators.

Solubility
Insoluble in water.

Notes
Air Sensitive & Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water, moisture, acids, brass, zinc, magnesium, aluminum, mercury, mercury oxides, acid chlorides, acid anhydrides, strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-50-3
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.11
MDL Number MFCD00008327
InChI Key GETQZCLCWQTVFV-UHFFFAOYSA-N
Synonym trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241
PubChem CID 1146
ChEBI CHEBI:18139
IUPAC Name N,N-dimethylmethanamine
SMILES CN(C)C

Specifications

Density 0.75
Flash Point −20°C (−4°F)
Refractive Index 1.3635
Linear Formula (CH3)3N
Quantity 250 mL
UN Number UN2924
Beilstein 956566
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,9710
Solubility Information Insoluble in water.
Formula Weight 59.11
Concentration or Composition (by Analyte or Components) 33% w/w in ethanol
Chemical Name or Material Trimethylamine
Show More Show Less
Hazard Category H225-H302+H332-H304-H314-H335-H336
Hazard Statement GHS H Statement
H225-H314-H302-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 200-875-0
TSCA Yes
Recommended Storage Ambient temperatures

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.