Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. T0465100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0465100G 100 g
T046525G 25 g
T0465500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0465100G Supplier TCI America Supplier No. T0465100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 593-81-7
Molecular Formula C3H10ClN
Molecular Weight (g/mol) 95.57
MDL Number MFCD00012478
InChI Key SZYJELPVAFJOGJ-UHFFFAOYSA-N
Synonym trimethylamine hydrochloride, trimethylammonium chloride, trimethylamine hcl, n,n-dimethylmethanamine hydrochloride, trimethylamine monohydrochloride, methanamine, n,n-dimethyl-, hydrochloride, trimethylaminehydrochloride, unii-uw38srm77k, uw38srm77k, trimethyl ammonium chloride
PubChem CID 10313079
ChEBI CHEBI:64700
IUPAC Name hydrogen trimethylamine chloride
SMILES [H+].[Cl-].CN(C)C

Specifications

Color White
Quantity 100 g
Formula Weight 95.57
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylamine Hydrochloride
RTECSNumber YH2700000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.