Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylhexamethylene Diisocyanate (2,2,4- and 2,4,4- mixture) 97.0+%, TCI America™
SDP

Catalog No. T1176100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1176100ML 100 mL
T117625ML 25 mL
T1176500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T1176100ML Supplier TCI America Supplier No. T1176100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28679-16-5
Molecular Formula C11H18N2O2
MDL Number MFCD00040422

Specifications

Color Yellow
Quantity 100 mL
UN Number 2328
Formula Weight 210.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylhexamethylene Diisocyanate (2,2,4- and 2,4,4- mixture)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.