Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylhydroquinone 98.0+%, TCI America™
SDP

Catalog No. T0477100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0477100G 100 g
T047725G 25 g
T0477500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0477100G Supplier TCI America Supplier No. T0477100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 700-13-0
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00002346
InChI Key AUFZRCJENRSRLY-UHFFFAOYSA-N
Synonym trimethylhydroquinone, 2,3,5-trimethylhydroquinone, 1,4-benzenediol, 2,3,5-trimethyl, pseudocumohydroquinone, psi-cumohydroquinone, 2,3,6-trimethylhydroquinone, 2,3,5-trimethyl-benzene-1,4-diol, hydroquinone, trimethyl, 2,3,5-trimethyl-1,4-benzenediol, .psi.-cumohydroquinone
PubChem CID 12785
IUPAC Name 2,3,5-trimethylbenzene-1,4-diol
SMILES CC1=CC(=C(C(=C1O)C)C)O

Specifications

Melting Point 169°C
Quantity 100 g
UN Number 3077
Formula Weight 152.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylhydroquinone
EINECSNumber (3)-0552&(4)-0057&(9)-0913
RTECSNumber MX7820000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.