Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylolpropane ethoxylate triacrylate

Catalog No. 4121981L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-219-81L 1 L
41-219-8250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 41-219-81L Supplier Sigma Aldrich Supplier No. 4121981L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethylolpropane ethoxylate triacrylate

Specifications

CAS 28961-43-5
Density 1.12 g/mL (at 25°C)
Boiling Point 157°C
Refractive Index n20/D 1.471 (literature)
Linear Formula [H2C=CHCO2(CH2CH2O)nCH2]3 CC2H5
Quantity 1 L
Synonym Trimethylolpropane ethoxylate (14/3 EO/OH) triacrylate
Molecular Weight (g/mol) 912
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.