Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylolpropane Trimethacrylate (stabilized with MEHQ) 90.0+%, TCI America™
SDP

Catalog No. T0912500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0912500ML 500 mL
T091225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0912500ML Supplier TCI America Supplier No. T0912500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3290-92-4
Molecular Formula C18H26O6
Molecular Weight (g/mol) 338.4
MDL Number MFCD00008588
InChI Key OKKRPWIIYQTPQF-UHFFFAOYSA-N
Synonym 2-Ethyl-2-hydroxymethyl-1,3-propanediol Trimethacrylate
PubChem CID 18689
ChEBI CHEBI:35029
IUPAC Name 2,2-bis(2-methylprop-2-enoyloxymethyl)butyl 2-methylprop-2-enoate
SMILES CCC(COC(=O)C(=C)C)(COC(=O)C(=C)C)COC(=O)C(=C)C

Specifications

Melting Point -14°C
Color Yellow
Boiling Point 155°C
Quantity 500 mL
Formula Weight 338.40
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylolpropane Trimethacrylate (stabilized with MEHQ)
EINECSNumber (2)-0769&(2)-1062
RTECSNumber TY6675000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.