Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylphenylammonium Bromide 98.0+%, TCI America™
SDP

Catalog No. P0243100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0243100G 100 g
P024325G 25 g
P0243500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0243100G Supplier TCI America Supplier No. P0243100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16056-11-4
Molecular Formula C9H14BrN
Molecular Weight (g/mol) 216.122
MDL Number MFCD00011788
InChI Key GNMJFQWRASXXMS-UHFFFAOYSA-M
Synonym trimethylphenylammonium bromide, phenyltrimethylammonium bromide, n,n,n-trimethylbenzenaminium bromide, benzenaminium, n,n,n-trimethyl-, bromide, n,n,n-trimethylanilinium bromide, phenyl trimethyl ammonium bromide, ammonium, phenyltrimethyl-, bromide, trimethyl phenyl azanium bromide, benzenaminium, n,n,n-trimethyl-, bromide 1:1, ammonium trimethylphenyl-, bromide
PubChem CID 27663
IUPAC Name trimethyl(phenyl)azanium;bromide
SMILES C[N+](C)(C)C1=CC=CC=C1.[Br-]

Specifications

Color White
Quantity 100 g
Formula Weight 216.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylphenylammonium Bromide
EINECSNumber (9)-1187
RTECSNumber BT2100000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.