Learn More
Sigma Organic Chemistry Trimethylsilyl acetate | 25G | 2754-27-0 | MFCD00008695
Supplier: Sigma Organic Chemistry 28445925G

CAS #: 2754-27-0
MDL #: MFCD00008695
Purity: >97 %
UNSPSC Code: 12352100
Molecular Weight: 132.23
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.