Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylsilyl acetate, 97%

Catalog No. p-7026181
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2203722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2203722 Supplier Thermo Scientific Chemicals Supplier No. B2203722
Only null left
Add to Cart

CAS: 2754-27-0 | C5H12O2Si | 132.234 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2754-27-0
Molecular Formula C5H12O2Si
Molecular Weight (g/mol) 132.234
MDL Number MFCD00008695
InChI Key QHUNJMXHQHHWQP-UHFFFAOYSA-N
Synonym acetoxytrimethylsilane, trimethylacetoxysilane, acetato trimethylsilane, acetic acid trimethylsilyl ester, silanol, trimethyl-, acetate, silanol, 1,1,1-trimethyl-, 1-acetate, tmsoac, o-trimethylsilylacetate, trimethylsilyl acetate
PubChem CID 75988
IUPAC Name trimethylsilyl acetate
SMILES CC(=O)O[Si](C)(C)C

Specifications

Melting Point -32°C
Density 0.884
Boiling Point 107°C
Flash Point 4°C (39°F)
Refractive Index 1.388
Linear Formula CH3CO2Si(CH3)3
Quantity 100 g
UN Number UN3272
Beilstein 1744463
Sensitivity Moisture sensitive
Formula Weight 132.24
Percent Purity 97%
Chemical Name or Material Trimethylsilyl acetate
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 220-404-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.