Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Trimethylsilyl)methylmagnesium chloride, 1.3M solution in THF, AcroSeal™

Catalog No. AC377468000
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle

CAS: 13170-43-9 | C4H11ClMgSi | 146.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13170-43-9, 109-99-9, 108-88-3
Molecular Formula C4H11ClMgSi
Molecular Weight (g/mol) 146.97
MDL Number MFCD00009916
InChI Key NAQATMJWCJCHOZ-UHFFFAOYSA-M
Synonym trimethylsilylmethylmagnesium chloride, tmsch2mgcl, trimethylsilyl methylmagnesium chloride, trimethylsilyl methyl magnesium chloride, trimethylsilyl methylmagnesium chloride solution, me3sich2mgcl, naqatmjwcjchoz-uhfffaoysa-m, chlorotrimethylsilylmethyl magnesium, trimethylsiylmethylmagnesium chloride
PubChem CID 2734900
SMILES C[Si](C)(C)C[Mg]Cl

Specifications

Color Colorless to Green
CAS Min % 18.37, 80.22
CAS Max % 19.78, 81.63
Linear Formula (CH3)3Si-CH2-MgCl
Concentration 1.2 to 1.5M
Fieser 16,118; 15,343; 12,540; 06,636; 05,724; 10,433
Density 1.0400g/mL
Solubility Information Solubility in water: reacts violently with water.
IUPAC Name magnesium;methanidyl(trimethyl)silane;chloride
Formula Weight 146.97
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.04
Quantity 800 mL
Flash Point −17°C
Physical Form Low Melting Solid
Chemical Name or Material (Trimethylsilyl)methylmagnesium chloride, 1.1M Solution in THF
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. May cause respiratory irritation. Causes severe skin burns and eye damage. Suspected of causing cancer. Reacts violently with water. May form explosive peroxides. Harmful if swallowed. May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.