Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylsilyl Trifluoromethanesulfonate 98.0+%, TCI America™
SDP

Catalog No. T0871250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0871250G 250 g
T08715G 5 g
T087125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0871250G Supplier TCI America Supplier No. T0871250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27607-77-8
Molecular Formula C4H9F3O3SSi
Molecular Weight (g/mol) 222.253
MDL Number MFCD00000406
InChI Key FTVLMFQEYACZNP-UHFFFAOYSA-N
Synonym trimethylsilyl triflate, tmsotf, trifluoromethanesulfonic acid trimethylsilyl ester, trimethylsilyl trifluoromethylsulphonate, unii-z84v0cbh9j, methanesulfonic acid, trifluoro-, trimethylsilyl ester, trimethylsilyl trifluoromethylsulfonate, trimethylsllytrifluoromethanesulphonate, trimethylsilyl trifluoromethanesulphonate, trimethylsilyltrifluoromethanesulphonate
PubChem CID 65367
IUPAC Name trimethylsilyl trifluoromethanesulfonate
SMILES C[Si](C)(C)OS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 140°C
Quantity 250 g
UN Number 2920
Formula Weight 222.25
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Trimethylsilyl Trifluoromethanesulfonate [Trimethylsilylating Agent]
EINECSNumber (2)-3617
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.