Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylsilylacetylene 98.0+%, TCI America™
SDP

Catalog No. T123925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T123925ML 25 mL
T12395ML 5 mL
T1239250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. T123925ML Supplier TCI America Supplier No. T123925ML
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 1066-54-2
Molecular Formula C5H10Si
Molecular Weight (g/mol) 98.22
MDL Number MFCD00008569
InChI Key CWMFRHBXRUITQE-UHFFFAOYSA-N
Synonym trimethylsilylacetylene, trimethylsilyl acetylene, silane, ethynyltrimethyl, ethynyl-trimethyl-silane, ethynyl trimethyl silane, tms acetylene, ethynyltrimethyl silane, trimethylsilyl-acetylene, tmsacetylene
PubChem CID 66111
IUPAC Name ethynyltrimethylsilane
SMILES C[Si](C)(C)C#C

Specifications

Color Colorless
Boiling Point 53°C
Quantity 25 mL
UN Number 1993
Formula Weight 98.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylsilylacetylene
EINECSNumber (2)-3779
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.