Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylsilylmethyl Acetate 98.0+%, TCI America™
SDP

Catalog No. A161325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A161325G 25 g
A16135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A161325G Supplier TCI America Supplier No. A161325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2917-65-9
Molecular Formula C6H14O2Si
Molecular Weight (g/mol) 146.26
MDL Number MFCD00009650
InChI Key TXZMVBJSLYBOMN-UHFFFAOYSA-N
Synonym trimethylsilyl methyl acetate, acetoxymethyltrimethylsilane, methanol, trimethylsilyl-, acetate, methanol, 1-trimethylsilyl-, 1-acetate, 2-trimethylsilyl methyl acetate, acmc-1ci3n, o-, acetoxymethyl trimethyl silane, txzmvbjslybomn-uhfffaoysa, acetic acid trimethylsilylmethyl ester
PubChem CID 76214
IUPAC Name (trimethylsilyl)methyl acetate
SMILES CC(=O)OC[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 137°C
Quantity 25 g
UN Number 3272
Formula Weight 146.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylsilylmethyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.