Learn More
Trimethyltin hydroxide, 98%
Description
Trimethyltin hydroxide is used in the synthesis of modified thymines, which is useful as inhibitors of RNase A. It is also used in the preparation of ketoheterocycle inhibitors of fatty acid amide inhibitors. It reacts with 2,2-disubstituted benzothiazolines to form trialkyltin derivatives of sulphur containing Schiff base complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 114°C to 118°C |
| Color | White |
| Odor | Pungent |
| Linear Formula | (CH3)3SnOH |
| Quantity | 5 g |
| UN Number | UN3146 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in water. |
| Formula Weight | 180.8 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H300+H310+H330 |
| Hazard Statement | GHS H Statement H300-H310-H330 Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. |
| Precautionary Statement | P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: III |
| RTECSNumber | WH8400000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.