Learn More
Apexbio Technology LLC Trimipramine (maleate),5g CAS# 521-78-8
SDP
Supplier:  Apexbio Technology LLC C57345000
Encompass_Preferred
Trimipramine (maleate) is a potent antagonist of histamine H1 receptor, serotonin 5-HT2A, and α1-adrenergic receptors [1].The histamine H1 receptor is widely expressed tissues, such as smooth muscles, vascular endothelial cells, heart, and the central nervous system. Histamine H1 receptor (H1R) antagonists are very effective drugs alleviating the symptoms of allergic reactions [2]. 5-HT2A receptor shows constitutive activity. Variations in5-HT2A receptor can produce profound alterations in cognitive states [3]. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0333