Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trinity Biotech Premier Diluent

Catalog No. 22140174
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
3800 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-140-174 3800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Irritating to eyes
Catalog No. 22-140-174 Supplier Trinity Biotech Supplier No. 01030097
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Irritating to eyes

Premier Diluent Reagent is designed for use with the Trinity Biotech Premier Hb9210 system for the quantitative measurement of the percentage of total glycated hemoglobins in whole blood

  • Premier Hb9210 system quantitatively measures hemoglobin A1c (HbA1c) in human capillary and venous whole blood
  • HbA1c is used for the monitoring of long-term glycemic control in individuals with diabetes mellitus
  • Reagents are performance validated to assure accuracy and precision with the Trinity Biotech assay and system
  • Reagent is ready for use

For in vitro diagnostic use only

Certifications

CE and FDA

Specifications

Chemical Name or Material Premier Hb9210 diluent reagent
Color Clear
For Use With (Application) For the quantitative measurement of the percentage of total Glycated Hemoglobins in whole Blood, for in-vitro diagnostic use
For Use With (Equipment) Trinity Biotech premier Hb9210 HbA1c analyzer
Physical Form Liquid
Quantity 3800 mL
Formulation Water 90 to 100%, TRITON X100 1 to 10%, Sodium Azide (Sodium Azide (as NaN3)) 0 to 0.5%
Certifications/Compliance FDA, CE, Canadian Licensed
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.