Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triolein 80.0+%, TCI America™
SDP

Catalog No. G0089100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0089100G 100 g
G008925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0089100G Supplier TCI America Supplier No. G0089100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 122-32-7
Molecular Formula C57H104O6
Molecular Weight (g/mol) 885.453
MDL Number MFCD00137563
InChI Key PHYFQTYBJUILEZ-BSCDBXJPSA-N
Synonym triolein, glycerol trioleate, glyceryl trioleate, oleic triglyceride, trioleoylglycerol, glycerol triolein, oleic acid triglyceride, trioleoylglyceride, olein, glycerin trioleate
PubChem CID 45253964
IUPAC Name [2-[(Z)-octadec-9-enoyl]oxy-3-[(E)-octadec-9-enoyl]oxypropyl] (E)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCCCCCCCC)OC(=O)CCCCCCCC=CCCCCCCCC

Specifications

Melting Point -4°C
Boiling Point 240°C
Quantity 100 g
Formula Weight 885.45
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Triolein
EINECSNumber (2)-0669
RTECSNumber RG1936500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.