Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIP13 Monoclonal Antibody (OTI2F5)
GREENER_CHOICE

Catalog No. PIMA527201
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527201 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527201 Supplier Invitrogen™ Supplier No. MA527201
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRIP13 Monoclonal Antibody for Western Blot, IHC (P)

Thyroid hormone receptors (TRs) are transcription factors that regulate the expression of specific genes in a hormone-dependent manner. TRIP13 (thyroid hormone receptor interactor 13), also called 16E1BP, is a transcription factor that interacts with the ligand binding domain of the thyroid receptor (TR) as well as a variety of target genes including human papilloma virus type 16 (HPV16) E1. Unlike most TRIP proteins which function only in the presence of hormones, TRIP13 does not require the presence of thyroid hormone to interact with TR. The association of TRIP13 with (HPV16) E1 suggests that TRIP13 may have tumor suppressor gene function. TRIP13 is a 432 amino acid protein with 2 different isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIP13
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2F5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TRIP13
Gene Accession No. Q15645
Gene Alias 16E1BP; 16E1-BP; 2410002G23Rik; D13Ertd328e; HPV16 E1 protein binding protein; HPV16 E1 protein-binding protein; human papillomavirus type 16 E1 protein-binding protein; Pachytene checkpoint protein 2 homolog; PCH2; Thyroid hormone receptor interactor 13; thyroid receptor interacting protein 13; thyroid receptor-interacting protein 13; TR-interacting protein 13; TRIP13; TRIP-13
Gene Symbols TRIP13
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 31-148 of TRIP13 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9319
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.