Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIP6 Mouse anti-Human, Mouse, Clone: 5D8C7, Proteintech
SDP

Catalog No. p-7213058 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6307 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6307 Supplier Proteintech Group Inc Supplier No. 602051IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene is a member of the zyxin family and encodes a protein with three LIM zinc-binding domains. This protein localizes to focal adhesion sites and along actin stress fibers. Recruitment of this protein to the plasma membrane occurs in a lysophosphatidic acid (LPA)-dependent manner and it regulates LPA-induced cell migration. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.

Specifications

Antigen TRIP6
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 5D8C7
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TRIP6
Gene Accession No. Q15654, Q9Z1Y4
Gene Alias OIP 1, OIP1, Opa interacting protein 1, TR interacting protein 6, TRIP 6, TRIP6, ZRP 1, Zyxin related protein 1
Gene Symbols TRIP6
Host Species Mouse
Immunogen TRIP6 Fusion Protein Ag15149
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22051, 7205
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.