Learn More
TARGETMOL CHEMICALS INC TRIPARANOL 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T262965MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Triparanol (NSC-65345) interferes with posttranslational modification of Hedgehog signaling molecules as well as the sterol sensing domain of its receptor PTCH1 leading to down-regulation of Hedgehog signaling. Triparanol suppresses human tumor growth and is an antilipemic agent with high ophthalmic toxicity. purity: 99%
Catalog No. 50-258-0709
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.