Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylbismuth, 98+%, Thermo Scientific Chemicals

Catalog No. AA1254522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1254522 100 g
AA1254506 5 g
AA1254514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1254522 Supplier Thermo Scientific Chemicals Supplier No. 01254522
Add to Cart

Triphenylbismuth, as an volatile organometallic used in the preparation of thin films of superconducting oxides by CVD. It is also added to the fuel of rocket as cruing agent. It acts as a solidifying catalyst for butyl hydroxyl propellants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Volatile organometallic used in the preparation of thin films of superconducting oxides by CVD
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-33-8
Molecular Formula C18H15Bi
Molecular Weight (g/mol) 440.298
MDL Number MFCD00014064
InChI Key ZHXAZZQXWJJBHA-UHFFFAOYSA-N
Synonym triphenylbismuth, triphenylbismuthine, bismuthine, triphenyl, aerotex, bismuth triphenyl, triphenyl bismuth, bismuth, triphenyl, trifenylbismutin, caswell no. 896, trifenylbismutin czech
PubChem CID 11774
IUPAC Name triphenylbismuthane
SMILES C1=CC=C(C=C1)[Bi](C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 76°C to 81°C
Color White
Density 1.585 g/mL
Boiling Point 242°C (14 mmHg)
Quantity 100 g
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in alcohol; Soluble in ether, acetone and chloroform; Insoluble in water
Formula Weight 440.3
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Triphenylbismuth
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 210-033-4
RTECSNumber EB2980000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.