Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylcarbenium tetrafluoroborate, 97%

Catalog No. AC172950500
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC172950500 50 g
AC172950050 5 g
AC172950250 25 g
AC172951000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC172950500 Supplier Thermo Scientific Chemicals Supplier No. 172950500

CAS: 341-02-6 | C19H15·BF4 | 330.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 341-02-6
MDL Number MFCD00013120
InChI Key VQXBOEYKSVVPSP-UHFFFAOYSA-N
Synonym triphenylcarbenium tetrafluoroborate, trityl tetrafluoroborate, tritylium tetrafluoroborate, triphenylmethylium tetrafluoroborate, diphenylmethylbenzene tetrafluoroborate, trityl fluoroborate, triphenylmethyl fluoroborate, triphenylcarbonium tetrafluoroborate, methylium, triphenyl-, tetrafluoroborate 1-, methylium, triphenyl-, tetrafluoroborate 1-1:1
PubChem CID 2723955
IUPAC Name diphenylmethylbenzene;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C1=CC=C(C=C1)[C+](C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 203°C to 213°C (decomposition)
Color Green to Ochre-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C19H15BF4
Linear Formula (C6H5)3CBF4
Quantity 50 g
Molecular Weight (g/mol) 330.133
Formula Weight 330.13
Percent Purity ≥96.0%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Triphenylcarbenium tetrafluoroborate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-433-3
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.