Learn More
eMolecules​ Triphenylmethane | 519-73-3 | MFCD00004763 | 25g
Supplier:  eMolecules​ 570996054
Encompass_Preferred
Ambeed | Triphenylmethane | 25g | 570996054 | A457629 | | 519-73-3 | MFCD00004763 | 244.337 | C19H16
If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool
Catalog No. 50-213-4735