Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylsilanol 98.0+%, TCI America™
SDP

Catalog No. T0945250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0945250G 250 g
T094525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0945250G Supplier TCI America Supplier No. T0945250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 791-31-1
Molecular Formula C18H16OSi
Molecular Weight (g/mol) 276.41
MDL Number MFCD00002102
InChI Key NLSXASIDNWDYMI-UHFFFAOYSA-N
Synonym triphenylsilanol, silanol, triphenyl, triphenylhydroxysilane, silane, hydroxytriphenyl, unii-azc8o4ttx4, silanol, 1,1,1-triphenyl, hydroxy triphenyl silane, si oh ph3, azc8o4ttx4
PubChem CID 69925
ChEBI CHEBI:35035
IUPAC Name hydroxy(triphenyl)silane
SMILES C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)O

Specifications

Melting Point 153°C
Color White
Quantity 250 g
Formula Weight 276.41
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylsilanol
EINECSNumber (3)-2638
RTECSNumber VV4325500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.