Learn More
Terumo BCT Inc COBE 2991 Triple Processing Set
Supplier:  Terumo BCT Inc 90901
Encompass
The triple processing set is most commonly used for bone marrow processing procedures on the COBE 2991 cell processor.
Catalog No. NC2102583