Learn More
Thermo Scientific™ Triple Sugar Iron Agar (TSI) Medium (Dehydrated)
Shop All Thermo Scientific ProductsDescription
Differentiate gram-negative enteric bacilli such as Enterobacteriaceae on the basis of dextrose, lactose and sucrose fermentation and hydrogen sulfide production using Thermo Scientific™ Triple Sugar Iron Agar (TSI) (Dehydrated).
TSI Agar is available as dehydrated culture medium or in ready-to-use prepared media tubes and in formats that meet international industrial guidelines for microbiological testing.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Certifications/Compliance | AOAC, BAM, COMPF, IDF, USDA, USP, SMWW |
| Color | Straw |
| Content And Storage | 20°C to 25°C |
| Description | Triple Sugar Iron Agar (TSI) Medium |
| Form | Powder |
| Manufacturing Origin | United States |
| Product Type | Agar |
| Fill Volume | 500 g |
| Packaging Type | Plastic bottle with red cap |
| Quantity | 1/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.