Learn More
Thermo Scientific™ TriPlus RSH Autosampler Fixed Needle Syringe, 10μL
Catalog No. 03375358 Shop All Thermo Scientific Products
Change view
Click to view available options
Needle Gauge:
23s G
Length (Metric) Needle:
57 mm
Catalog No. Needle Gauge Length (Metric) Needle
03375358 23s G 57 mm
1 options
Catalog No. 03375358 Supplier Thermo Scientific™ Supplier No. 365D0271
Add to Cart
Add to Cart
Thermo Scientific™ TriPlus RSH Autosampler Fixed Needle Syringes are accurate and reproducible over a range of volumes.
  • Borosilicate glass with metal flanges
  • Needles and plungers are stainless steel
  • Field-proven syringe design
TRUSTED_SUSTAINABILITY
Specifications
Description Merlin Valve Injector, PTV and Split
Needle Gauge 23s G
Product Type GC Autosampler Syringe, Fixed Needle
Volume (Metric) 10 μL
Length (Metric) Needle 57 mm
Needle Point Style Cone
For Use With (Equipment) TriPlus RSH Autosampler and TriPlus 100LS Autosampler