Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tripropyl Orthoformate 98.0+%, TCI America™
SDP

Catalog No. O0259500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0259500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O0259500ML Supplier TCI America Supplier No. O0259500ML

Chemical Identifiers

CAS 621-76-1
Molecular Formula C10H22O3
Molecular Weight (g/mol) 190.283
MDL Number MFCD00015214
InChI Key RWNXXQFJBALKAX-UHFFFAOYSA-N
Synonym tripropyl orthoformate, tri-n-propyl orthoformate, orthoformic acid tripropyl ester, 1-dipropoxymethoxy propane, unii-z8f0df6af9, propane, 1,1',1-methylidynetris oxy tris, z8f0df6af9, tripropoxymethane, tripropylorthoformate
PubChem CID 69311
IUPAC Name 1-(dipropoxymethoxy)propane
SMILES CCCOC(OCCC)OCCC

Specifications

Color Yellow
Boiling Point 198°C
Quantity 500 mL
Formula Weight 190.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tripropyl Orthoformate
EINECSNumber (2)-3932
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.