Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Triptolide

Catalog No. AC466841000
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466841000 100 mg
AC466840250 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466841000 Supplier Thermo Scientific Chemicals Supplier No. 466841000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 38748-32-2 | C20H24O6, C20H24O6 | 360.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38748-32-2
Molecular Formula C20H24O6

Specifications

Quantity 100 mg
Solubility Solubility aq. soln.: .. Other solubilities: soluble in dmso, ethanol
Melting Point 226.0°C to 227.0°C
Formula Weight 360.41
Chemical Name or Material Triptolide
What storage conditions should I use for Triptolide?

We recommend storing Triptolide in powder form at -20°C, desiccated. Once in solution, it should be stored at -20°C and used within 1 month to prevent loss of potency. We suggest you make aliquots of the stock solution to avoid multiple freeze/thaw cycles.

How can I prepare a 10 mM stock solution of Triptolide?

To prepare a 10 mM stock of Triptolide, we recommend that you reconstitute 5 mg of powder in 1.38 mL of DMSO.

Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Store locked up.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.