Learn More
ILEX LIFE SCIENCES LLC Triptorelin Acetate (GnRH agonist), Synthetic Peptide, 10 mg
SDP
Supplier:  ILEX LIFE SCIENCES LLC A223810
Encompass_Preferred
Triptorelin is a synthetic analogue of gonadorelin (GnRH). The only change between GnRH and Triptorelin is the introduction of a residue of D-Triptophane instead of a glycine in position 6. As a result of the substitution of the 6th amino acid residue in the native molecule, the agonistic effect is more pronounced and the plasma half-life prolonged. Triptorelin controls release of Luteinizing Hormone (LH) and Follicle Stimulating Hormone (FSH) and also levels of testosterone and estrogen. CAS NO.: 57773-63-4. MOLECULAR FORMULA: C64H82N18O13. AMINO ACID SEQUENCE: Pyr-His-Trp-Ser-Tyr-D-Trp-Leu-Arg-Pro-Gly-NH2. TYPE: Synthetic peptide. PURITY: Greater than 98.0%. FORMULATION: Sterile-filtered white lyophilized (freeze-dried) powder. The peptide (1 mg/ml) was lyophilized with no additives. OTHER NAMES: Gonadotropin-releasing hormone (GnRH) superagonist, luteinizing hormone-releasing hormone agonist, luteinising hormone-releasing hormone agonist, GnRH agonist, LHRH agonist.
Catalog No. 50-233-9560
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.