Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triptycene-9,10-dicarboxaldehyde 96.0+%, TCI America™
SDP

Catalog No. T31461G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31461G 1 g
T3146200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31461G Supplier TCI America Supplier No. T31461G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 467429-73-8
Molecular Formula C22H14O2
Molecular Weight (g/mol) 310.35
MDL Number MFCD29472555
InChI Key HTLPGDCICALQIK-UHFFFAOYSA-N
Synonym 9,10-Diformyltriptycene
PubChem CID 131637027
IUPAC Name pentacyclo[6.6.6.0²,⁷.0⁹,¹⁴.0¹⁵,²⁰]icosa-2,4,6,9,11,13,15,17,19-nonaene-1,8-dicarbaldehyde
SMILES O=CC12C3=CC=CC=C3C(C=O)(C3=CC=CC=C13)C1=CC=CC=C21

Specifications

Color Yellow
Quantity 1 g
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Triptycene-9,10-dicarboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.