Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tripyrrolidinophosphine Oxide 98.0+%, TCI America™
SDP

Catalog No. T295225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T295225G 25 g
T29525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T295225G Supplier TCI America Supplier No. T295225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6415-07-2
Molecular Formula C12H24N3OP
Molecular Weight (g/mol) 257.32
MDL Number MFCD00014095
InChI Key GQOGEHIVQIMJMO-UHFFFAOYSA-N
Synonym Tris(N,N-tetramethylene)phosphoric Triamide
PubChem CID 80866
IUPAC Name 1-dipyrrolidin-1-ylphosphorylpyrrolidine
SMILES C1CCN(C1)P(=O)(N2CCCC2)N3CCCC3

Specifications

Color Yellow
Boiling Point 133°C
Quantity 25 g
Formula Weight 257.32
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tripyrrolidinophosphine Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.