Learn More
Tris[2-(2-methoxyethoxy)ethyl]amine 90.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Specifications
Specifications
| Boiling Point | 171°C |
| Quantity | 100 g |
| UN Number | 2735 |
| Formula Weight | 323.43 |
| Percent Purity | ≥90.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Tris[2-(2-methoxyethoxy)ethyl]amine |
Safety and Handling
| TSCA | Yes |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.