Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2-chloroethyl) Phosphate 93.0+%, TCI America™
SDP

Catalog No. T3447100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3447100G 100 g
T344725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3447100G Supplier TCI America Supplier No. T3447100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 115-96-8
Molecular Formula C6H12Cl3O4P
Molecular Weight (g/mol) 285.482
MDL Number MFCD00000967
InChI Key HQUQLFOMPYWACS-UHFFFAOYSA-N
Synonym tris 2-chloroethyl phosphate, celluflex, fyrol cef, disflamoll tca, celluflex cef, niax 3cf, tris chloroethyl phosphate, trichlorethyl phosphate, 2-chloroethanol phosphate
PubChem CID 8295
ChEBI CHEBI:35037
IUPAC Name tris(2-chloroethyl) phosphate
SMILES C(CCl)OP(=O)(OCCCl)OCCCl

Specifications

Color Yellow
Boiling Point 146°C
Quantity 100 g
Formula Weight 285.48
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris(2-chloroethyl) Phosphate
EINECSNumber (2)-1941
RTECSNumber KK2450000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.