Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2-phenylpyridinato)iridium(III) (purified by sublimation) 95.0+%, TCI America™
SDP

Catalog No. T1946200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1946200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1946200MG Supplier TCI America Supplier No. T1946200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Purified by sublimation, this organometallic catalyst is designed for photoredox-catalyzed chemical synthesis.

Chemical Identifiers

CAS 94928-86-6
Molecular Formula C33H24IrN3
Molecular Weight (g/mol) 654.793
MDL Number MFCD04039766
InChI Key NSABRUJKERBGOU-UHFFFAOYSA-N
Synonym Ir(ppy)3
PubChem CID 59117881
IUPAC Name iridium(3+);2-phenylpyridine
SMILES C1=CC=C([C-]=C1)C2=CC=CC=N2.C1=CC=C([C-]=C1)C2=CC=CC=N2.C1=CC=C([C-]=C1)C2=CC=CC=N2.[Ir+3]

Specifications

Melting Point 451°C
Quantity 200 mg
Formula Weight 654.79
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2-phenylpyridinato)iridium(III) (purified by sublimation)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.