accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 94928-86-6

IUPAC Name: 2-phenylpyridine
Synonyms: 2-phenylpyridine
Molecular Weight (g/mol): 155.2
Molecular Formula: C11H9N
InChi Key: VQGHOUODWALEFC-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC=CC=N1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)