Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(4-chlorophenyl)phosphine, 97%

Catalog No. AAA1248706
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1248706 5 g
AAA1248703 1 g
AAA1248714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1248706 Supplier Thermo Scientific Chemicals Supplier No. A1248706
Add to Cart

CAS: 1159-54-2 | C18H12Cl3P | 365.618 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1159-54-2
Molecular Formula C18H12Cl3P
Molecular Weight (g/mol) 365.618
MDL Number MFCD00013639
InChI Key IQKSLJOIKWOGIZ-UHFFFAOYSA-N
Synonym tris 4-chlorophenyl phosphine, tri p-chlorophenyl phosphine, tris p-chlorophenyl phosphine, tris 4-chlorophenyl phosphane, tri-4-chlorophenyl phosphine, phosphine, tris 4-chlorophenyl, tri 4-chlorophenyl phosphine, acmc-20ajbt, pubchem6454
PubChem CID 70874
IUPAC Name tris(4-chlorophenyl)phosphane
SMILES C1=CC(=CC=C1P(C2=CC=C(C=C2)Cl)C3=CC=C(C=C3)Cl)Cl

Specifications

Melting Point 102°C to 103°C
Quantity 5 g
Beilstein 751458
Sensitivity Air sensitive
Formula Weight 365.63
Percent Purity 97%
Chemical Name or Material Tris(4-chlorophenyl)phosphine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-596-7
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.