Learn More
Tris(4-methoxyphenyl)phosphine, 98%
Description
It is employed in Suzuki reaction. It is also used in the kinetic study of the effect of catalysts on the curing of biphenyl epoxy resin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is employed in Suzuki reaction. It is also used in the kinetic study of the effect of catalysts on the curing of biphenyl epoxy resin.
Solubility
Insoluble in water.
Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 130°C to 132°C |
| Quantity | 5 g |
| Beilstein | 2815911 |
| Sensitivity | Air Sensitive |
| Solubility Information | Insoluble in water. |
| Formula Weight | 352.37 |
| Percent Purity | 98% |
| Chemical Name or Material | Tris(4-methoxyphenyl)phosphine |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 212-723-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.