Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(4-methylphenyl)phosphine Oxide 98.0+%, TCI America™
SDP

Catalog No. T09951G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T09951G 1 g
T09955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T09951G Supplier TCI America Supplier No. T09951G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 797-70-6
Molecular Formula C21H21OP
Molecular Weight (g/mol) 320.37
MDL Number MFCD00059321
InChI Key SPKBYIYIZQARNX-UHFFFAOYSA-N
Synonym Tri-p-tolylphosphine Oxide
PubChem CID 120383
IUPAC Name 1-[bis(4-methylphenyl)phosphoroso]-4-methylbenzene
SMILES CC1=CC=C(C=C1)P(=O)(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1

Specifications

Melting Point 145°C
Color White
Quantity 1 g
Formula Weight 320.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tris(4-methylphenyl)phosphine Oxide
RTECSNumber SZ2000000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.