Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris, 99.8 to 100.1% (Dry Basis), Molecular Biology Grade, Ultrapure

Catalog No. AAJ75825A1
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
10 kg
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ75825A1 1 kg Plastic bottle
AAJ7582536 500 g Plastic bottle
AAJ75825A7 5 kg Plastic bottle
AAJ75825A9 10 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ75825A1 Supplier Thermo Scientific Supplier No. J75825A1
Add to Cart

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym Tris(hydroxymethyl)amino methane
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Melting Point 171°C to 172°C
Boiling Point 219°C to 220°C (10 mmHg)
Physical Form Crystalline Powder
Quantity 1 kg
Linear Formula NH2C(CH2OH)3
Packaging Plastic bottle
Beilstein 741883
Formula Weight 121.14
Grade Molecular Biology
Chemical Name or Material Tris
EINECSNumber 201-064-4
RTECSNumber TY2900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.