Learn More
United States Biological Corporation Tris Base Ultrapure, 77-86-1, MFCD00004679, 5kg
Catalog No. NC0622393 Shop All United States Biological Corporation Products
Encompass_Preferred
Molecular Formula C₄H₁₁NO₃, Purity ≥99.9%, Molecular Biology Grade, Molecular Weight 121.14, Tris is one of the most widely used buffers in molecular biology and cell culture due to its low toxicity, stability and buffering capacity.
Catalog No. NC0622393 Supplier United States Biological Corporation Supplier No. T86005KG
Molecular Formula C₄H₁₁NO₃, Purity ≥99.9%, Molecular Biology Grade, Molecular Weight 121.14, Tris is one of the most widely used buffers in molecular biology and cell culture due to its low toxicity, stability and buffering capacity.